Product Details

SNP ID
rs200770058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3493865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGCCTCTCCAGGGCCCCAGAGC[A/G]GGTTCTCTTGCACAAGGGACACATG
Phenotype
MIM: 616787 MIM: 614246
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf90 PubMed Links

Gene Details

Gene
C16orf90
Gene Name
chromosome 16 open reading frame 90
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080524.1 589 Missense Mutation CGC,TGC R175C NP_001073993.1
XM_005255502.3 589 Missense Mutation CGC,TGC R190C XP_005255559.1
Gene
CLUAP1
Gene Name
clusterin associated protein 1
There are no transcripts associated with this gene.

Gene
MIR6126
Gene Name
microRNA 6126
There are no transcripts associated with this gene.

Gene
NAA60
Gene Name
N(alpha)-acetyltransferase 60, NatF catalytic subunit
There are no transcripts associated with this gene.

View Full Product Details