Product Details

SNP ID
rs201126167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30709514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCCCTTTCACATCTGTGGCAGG[G/T]GGTGCAATTCAAGCAACAGTCCCGG
Phenotype
MIM: 611421
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SNORA30 PubMed Links

Gene Details

Gene
SNORA30
Gene Name
small nucleolar RNA, H/ACA box 30
There are no transcripts associated with this gene.

Gene
SRCAP
Gene Name
Snf2-related CREBBP activator protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006662.2 1020 Missense Mutation GGG,GTG G212V NP_006653.2

View Full Product Details