Product Details
- SNP ID
-
rs201257386
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:84009799 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGCTGCCCATCAGAACATCTCCCA[A/G]ACCGCTGCCGCCGTGCTCTGCCCAA
- Phenotype
-
MIM: 615585
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NECAB2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7196330] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NECAB2
- Gene Name
- N-terminal EF-hand calcium binding protein 2
There are no transcripts associated with this gene.
- Gene
- SLC38A8
- Gene Name
- solute carrier family 38 member 8
View Full Product Details