Product Details

SNP ID
rs201323582
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:72076796 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACCAATACGATTGCATAACGCA[A/T]TATGAAGGCAGCACATGCCCCAAAT
Phenotype
MIM: 140210
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HPR PubMed Links

Gene Details

Gene
HPR
Gene Name
haptoglobin-related protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020995.3 792 Missense Mutation CAA,CAT Q254H NP_066275.3
Gene
TXNL4B
Gene Name
thioredoxin like 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142317.1 792 Intron NP_001135789.1
NM_001142318.1 792 Intron NP_001135790.1
NM_001324354.1 792 Intron NP_001311283.1
NM_001324355.1 792 Intron NP_001311284.1
NM_017853.2 792 Intron NP_060323.1
XM_017023377.1 792 Intron XP_016878866.1

View Full Product Details