Product Details

SNP ID
rs200313433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71448477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCTGATGCCTAGTTAGTTTGGC[A/G]TTGATACTGAAGGCTTTCCCACATT
Phenotype
MIM: 194527
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF23 PubMed Links

Gene Details

Gene
ZNF23
Gene Name
zinc finger protein 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304492.1 2371 Intron NP_001291421.1
NM_001304493.1 2371 Silent Mutation AAC,AAT N458N NP_001291422.1
NM_001304494.1 2371 Intron NP_001291423.1
NM_145911.2 2371 Silent Mutation AAC,AAT N516N NP_666016.1

View Full Product Details