Product Details

SNP ID
rs200970733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57516331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCCAGCTCCTTCTTGAGCTCCT[C/T]CACCCGCAGCCGCAGCTTCTTCACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC102A PubMed Links

Gene Details

Gene
CCDC102A
Gene Name
coiled-coil domain containing 102A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033212.3 1594 Missense Mutation AAG,GAG K461E NP_149989.2
XM_011523469.2 1594 Missense Mutation AAG,GAG K461E XP_011521771.1

View Full Product Details