Product Details
- SNP ID
-
rs199532631
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:564703 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCACCGGCTCCTCTGTGATGCTGT[C/G]GCCTGAGGACGGGGATCCAGGCGGC
- Phenotype
-
MIM: 605754
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC107987233
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs60243750] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC107987233
- Gene Name
- uncharacterized LOC107987233
There are no transcripts associated with this gene.
- Gene
- NHLRC4
- Gene Name
- NHL repeat containing 4
There are no transcripts associated with this gene.
- Gene
- PIGQ
- Gene Name
- phosphatidylinositol glycan anchor biosynthesis class Q
There are no transcripts associated with this gene.
- Gene
- PRR35
- Gene Name
- proline rich 35
View Full Product Details