Product Details

SNP ID
rs200219778
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56867121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTTTGACAGCCGGCCCAGCCAC[C/G]AGATGACTGATGGGCTGGTGGAGGG
Phenotype
MIM: 613395 MIM: 600968
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
MIR138-2 PubMed Links

Gene Details

Gene
MIR138-2
Gene Name
microRNA 138-2
There are no transcripts associated with this gene.

Gene
SLC12A3
Gene Name
solute carrier family 12 member 3
There are no transcripts associated with this gene.

View Full Product Details