Product Details
- SNP ID
-
rs201511674
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:3383073 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATGGGCTCTCCCCAGTGTGGATGC[A/G]CCGGTGAGCACTGAACTGGGAACTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZSCAN32
PubMed Links
Gene Details
- Gene
- ZSCAN32
- Gene Name
- zinc finger and SCAN domain containing 32
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001284527.1 |
2136 |
Missense Mutation |
CGC,TGC |
R625C |
NP_001271456.1 |
NM_001284528.1 |
2136 |
Missense Mutation |
CGC,TGC |
R336C |
NP_001271457.1 |
NM_001284529.1 |
2136 |
Missense Mutation |
CGC,TGC |
R336C |
NP_001271458.1 |
NM_001324340.1 |
2136 |
Missense Mutation |
CGC,TGC |
R413C |
NP_001311269.1 |
NM_001324341.1 |
2136 |
Missense Mutation |
CGC,TGC |
R413C |
NP_001311270.1 |
NM_001324342.1 |
2136 |
Missense Mutation |
CGC,TGC |
R413C |
NP_001311271.1 |
NM_001324343.1 |
2136 |
Missense Mutation |
CGC,TGC |
R492C |
NP_001311272.1 |
NM_001324344.1 |
2136 |
Missense Mutation |
CGC,TGC |
R413C |
NP_001311273.1 |
NM_001324345.1 |
2136 |
Missense Mutation |
CGC,TGC |
R413C |
NP_001311274.1 |
NM_001324346.1 |
2136 |
Missense Mutation |
CGC,TGC |
R552C |
NP_001311275.1 |
NM_017810.3 |
2136 |
Missense Mutation |
CGC,TGC |
R413C |
NP_060280.2 |
XM_005255403.2 |
2136 |
Missense Mutation |
CGC,TGC |
R538C |
XP_005255460.1 |
XM_011522555.2 |
2136 |
Missense Mutation |
CGC,TGC |
R625C |
XP_011520857.1 |
XM_017023371.1 |
2136 |
Missense Mutation |
CGC,TGC |
R625C |
XP_016878860.1 |
XM_017023372.1 |
2136 |
Missense Mutation |
CGC,TGC |
R538C |
XP_016878861.1 |
XM_017023373.1 |
2136 |
Missense Mutation |
CGC,TGC |
R538C |
XP_016878862.1 |
View Full Product Details