Product Details

SNP ID
rs200549368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:74999884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCGGGGGGGTGCCCTTTGGCCT[C/G]TACACCCCCGCCTCCCGGGGCACCG
Phenotype
MIM: 612060
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNRF1 PubMed Links

Gene Details

Gene
ZNRF1
Gene Name
zinc and ring finger 1, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032268.4 842 Silent Mutation CTC,CTG L71L NP_115644.1
XM_011523392.1 842 Silent Mutation CTC,CTG L71L XP_011521694.1
XM_017023793.1 842 Silent Mutation CTC,CTG L71L XP_016879282.1
XM_017023794.1 842 Silent Mutation CTC,CTG L71L XP_016879283.1

View Full Product Details