Product Details

SNP ID
rs200813960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31427831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACGTGGCTATTGTGGAACAGAGT[A/G]TGGTTGCCGTCCCCCCAGGGGTAGG
Phenotype
MIM: 602009 MIM: 602453
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COX6A2 PubMed Links

Gene Details

Gene
COX6A2
Gene Name
cytochrome c oxidase subunit 6A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005205.3 340 Silent Mutation CAC,CAT H79H NP_005196.1
Gene
ITGAD
Gene Name
integrin subunit alpha D
There are no transcripts associated with this gene.

Gene
ZNF843
Gene Name
zinc finger protein 843
There are no transcripts associated with this gene.

View Full Product Details