Product Details

SNP ID
rs201364860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3089529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGCCCTCACCGCACTCGCTGCA[G/T]CGGCAGGGCTTCTCGCCCGTGTGGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZSCAN10 PubMed Links

Gene Details

Gene
ZSCAN10
Gene Name
zinc finger and SCAN domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282415.1 1832 Silent Mutation CGA,CGC R241R NP_001269344.1
NM_001282416.1 1832 Silent Mutation CGA,CGC R498R NP_001269345.1
NM_032805.2 1832 Silent Mutation CGA,CGC R635R NP_116194.2
XM_017023791.1 1832 Silent Mutation CGA,CGC R635R XP_016879280.1
XM_017023792.1 1832 Silent Mutation CGA,CGC R603R XP_016879281.1

View Full Product Details