Product Details

SNP ID
rs200257202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:55567253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAAGCAGTATGACAGGGACCATT[A/C]TGGGTCTCTGGGAAGTTCTCAGCTG
Phenotype
MIM: 616767 MIM: 612040
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CAPNS2 PubMed Links

Gene Details

Gene
CAPNS2
Gene Name
calpain small subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032330.1 582 Missense Mutation TAT,TCT Y166S NP_115706.1
Gene
LPCAT2
Gene Name
lysophosphatidylcholine acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017839.4 582 Intron NP_060309.2
XM_005256006.2 582 Intron XP_005256063.1
XM_011523169.2 582 Intron XP_011521471.1
XM_017023376.1 582 Intron XP_016878865.1

View Full Product Details