Product Details

SNP ID
rs200324271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4425910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCGCGGTGCTCCACACGCTGGT[G/T]GCTGGTGGTTGTGGGGACCCCGCGG
Phenotype
MIM: 611668 MIM: 608382
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CORO7 PubMed Links

Gene Details

Gene
CORO7
Gene Name
coronin 7
There are no transcripts associated with this gene.

Gene
CORO7-PAM16
Gene Name
CORO7-PAM16 readthrough
There are no transcripts associated with this gene.

Gene
DNAJA3
Gene Name
DnaJ heat shock protein family (Hsp40) member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135110.2 106 Missense Mutation TGG,TTG W10L NP_001128582.1
NM_001286516.1 106 UTR 5 NP_001273445.1
NM_005147.5 106 Missense Mutation TGG,TTG W10L NP_005138.3

View Full Product Details