Product Details

SNP ID
rs202019779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:25239869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCTTATGTGGGTTCTCAGGGC[A/G]ATACAGAGATGGAGGGTGTGGCAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZKSCAN2 PubMed Links

Gene Details

Gene
ZKSCAN2
Gene Name
zinc finger with KRAB and SCAN domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012981.4 3258 Missense Mutation CGC,TGC R951C NP_001012999.3
XM_017023200.1 3258 Missense Mutation CGC,TGC R747C XP_016878689.1

View Full Product Details