Product Details

SNP ID
rs202155490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28884627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATCCTCGCCATCAAATCCACCAC[A/G]CTGCGGGTTGACCAGTCCATCCTGA
Phenotype
MIM: 108730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP2A1 PubMed Links

Gene Details

Gene
ATP2A1
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286075.1 700 Silent Mutation ACA,ACG T47T NP_001273004.1
NM_004320.4 700 Silent Mutation ACA,ACG T172T NP_004311.1
NM_173201.3 700 Silent Mutation ACA,ACG T172T NP_775293.1
Gene
ATP2A1-AS1
Gene Name
ATP2A1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details