Product Details

SNP ID
rs201930048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30086268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCACATCCAGATAGCCCCCAGGC[A/G]CGGTGTATGGTAGAGGGAAGGGGCC
Phenotype
MIM: 602035 MIM: 602427 MIM: 609724
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP4C PubMed Links

Gene Details

Gene
PPP4C
Gene Name
protein phosphatase 4 catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303503.1 1353 Intron NP_001290432.1
NM_001303504.1 1353 Intron NP_001290433.1
NM_001303506.1 1353 Intron NP_001290435.1
NM_001303507.1 1353 Intron NP_001290436.1
NM_002720.2 1353 Intron NP_002711.1
XM_006721061.3 1353 Intron XP_006721124.1
XM_017023403.1 1353 Intron XP_016878892.1
XM_017023404.1 1353 Intron XP_016878893.1
Gene
TBX6
Gene Name
T-box 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004608.3 1353 Missense Mutation GCG,GTG A423V NP_004599.2
XM_005255523.2 1353 Missense Mutation GCG,GTG A423V XP_005255580.1
XM_011545926.2 1353 Missense Mutation GCG,GTG A423V XP_011544228.1
XM_017023614.1 1353 Missense Mutation GCG,GTG A423V XP_016879103.1
Gene
YPEL3
Gene Name
yippee like 3
There are no transcripts associated with this gene.

View Full Product Details