Product Details

SNP ID
rs202103494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84222357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGTGTTTTGGAGGTGGCGGAGGC[A/G]GGCCTGAAGCTGCTCCTGCTCCTTC
Phenotype
MIM: 607603
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNG4 PubMed Links

Gene Details

Gene
KCNG4
Gene Name
potassium voltage-gated channel modifier subfamily G member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172347.2 1541 Missense Mutation CGC,TGC R474C NP_758857.1

View Full Product Details