Product Details

SNP ID
rs202038449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:81041364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTTTCCTTTTGCAGGATGGCAT[A/G]GTCAATCCAACAATAAGAAAAGATT
Phenotype
MIM: 614693 MIM: 611509
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATMIN PubMed Links

Gene Details

Gene
ATMIN
Gene Name
ATM interactor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300728.1 303 UTR 5 NP_001287657.1
NM_015251.2 303 Missense Mutation ATA,ATG I115M NP_056066.2
Gene
CENPN
Gene Name
centromere protein N
There are no transcripts associated with this gene.

View Full Product Details