Product Details

SNP ID
rs201718271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30067252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCCAGGAACTTGCTACTACCAGC[A/T]CCATGCCCTACCAATATCCAGCACT
Phenotype
MIM: 103850 MIM: 602035
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ALDOA PubMed Links

Gene Details

Gene
ALDOA
Gene Name
aldolase, fructose-bisphosphate A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000034.3 1085 UTR 5 NP_000025.1
NM_001127617.2 1085 UTR 5 NP_001121089.1
NM_001243177.1 1085 Missense Mutation ACC,TCC T54S NP_001230106.1
NM_184041.2 1085 UTR 5 NP_908930.1
NM_184043.2 1085 UTR 5 NP_908932.1
XM_011545768.2 1085 UTR 5 XP_011544070.1
Gene
PPP4C
Gene Name
protein phosphatase 4 catalytic subunit
There are no transcripts associated with this gene.

View Full Product Details