Product Details

SNP ID
rs199937543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:56834963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGCACCGCAGGGACATCTTCCG[C/G]AAGAGCAAGCACGGGTGGCGCGACA
Phenotype
MIM: 601440
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C17orf67 PubMed Links

Gene Details

Gene
C17orf67
Gene Name
chromosome 17 open reading frame 67
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085430.2 348 Intron NP_001078899.2
XM_011524735.2 348 Intron XP_011523037.1
Gene
DGKE
Gene Name
diacylglycerol kinase epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003647.2 348 Silent Mutation CGC,CGG R56R NP_003638.1
XM_011525394.2 348 Silent Mutation CGC,CGG R74R XP_011523696.1
XM_011525395.2 348 Silent Mutation CGC,CGG R74R XP_011523697.1
XM_011525396.2 348 Silent Mutation CGC,CGG R74R XP_011523698.1
XM_017025243.1 348 Silent Mutation CGC,CGG R56R XP_016880732.1
XM_017025244.1 348 Silent Mutation CGC,CGG R74R XP_016880733.1
XM_017025245.1 348 Silent Mutation CGC,CGG R74R XP_016880734.1

View Full Product Details