Product Details

SNP ID
rs200206773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30385193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCAGGAGATGAAGACGAGACTTT[C/G]AAAGATGGAATCACAAACGGCGCAC
Phenotype
MIM: 603102
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CPD PubMed Links

Gene Details

Gene
CPD
Gene Name
carboxypeptidase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199775.1 370 Missense Mutation TTC,TTG F70L NP_001186704.1
NM_001304.4 370 Missense Mutation TTC,TTG F317L NP_001295.2

View Full Product Details