Product Details

SNP ID
rs200381289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32334825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTGATTCTTTAACTTCAACAGG[A/G]GTGTAACTCAAAGCCTATGAGAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf75 PubMed Links

Gene Details

Gene
C17orf75
Gene Name
chromosome 17 open reading frame 75
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022344.3 772 Silent Mutation ACC,ACT T228T NP_071739.2
XM_005258022.3 772 Silent Mutation ACC,ACT T220T XP_005258079.1
XM_017024940.1 772 Silent Mutation ACC,ACT T49T XP_016880429.1
XM_017024941.1 772 Silent Mutation ACC,ACT T49T XP_016880430.1

View Full Product Details