Product Details
- SNP ID
-
rs199561996
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:74695842 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGGTCTGCATAGCAGAGGTCGCC[C/T]TCCAGGGGCTGCAGTACCTGGGACA
- Phenotype
-
MIM: 609807
MIM: 609956
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD300LF
PubMed Links
Gene Details
- Gene
- CD300LF
- Gene Name
- CD300 molecule like family member f
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001289082.1 |
810 |
Missense Mutation |
AGG,GGG |
R166G |
NP_001276011.1 |
NM_001289083.1 |
810 |
Missense Mutation |
AGG,GGG |
R216G |
NP_001276012.1 |
NM_001289084.1 |
810 |
Silent Mutation |
GAA,GAG |
E215E |
NP_001276013.1 |
NM_001289085.1 |
810 |
Silent Mutation |
GAA,GAG |
E203E |
NP_001276014.1 |
NM_001289086.1 |
810 |
Missense Mutation |
AGG,GGG |
R211G |
NP_001276015.1 |
NM_001289087.1 |
810 |
Missense Mutation |
AGG,GGG |
R193G |
NP_001276016.1 |
NM_139018.4 |
810 |
Silent Mutation |
GAA,GAG |
E200E |
NP_620587.2 |
XM_017024212.1 |
810 |
Silent Mutation |
GAA,GAG |
E219E |
XP_016879701.1 |
XM_017024213.1 |
810 |
Missense Mutation |
AGG,GGG |
R238G |
XP_016879702.1 |
XM_017024214.1 |
810 |
Missense Mutation |
AGG,GGG |
R235G |
XP_016879703.1 |
XM_017024215.1 |
810 |
Missense Mutation |
AGG,GGG |
R219G |
XP_016879704.1 |
XM_017024216.1 |
810 |
Missense Mutation |
AGG,GGG |
R212G |
XP_016879705.1 |
XM_017024217.1 |
810 |
Missense Mutation |
AGG,GGG |
R208G |
XP_016879706.1 |
- Gene
- RAB37
- Gene Name
- RAB37, member RAS oncogene family
View Full Product Details