Product Details

SNP ID
rs199533985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82048759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCGGGGCGCCCTGTTTCTGCCT[A/G]GGACACCAGTCCGTGTCTGGGTACA
Phenotype
MIM: 601934 MIM: 602578
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUS1L PubMed Links

Gene Details

Gene
DUS1L
Gene Name
dihydrouridine synthase 1 like
There are no transcripts associated with this gene.

Gene
GPS1
Gene Name
G protein pathway suppressor 1
There are no transcripts associated with this gene.

Gene
RFNG
Gene Name
RFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002917.1 971 Silent Mutation CCC,CCT P321P NP_002908.1
XM_011523587.2 971 Silent Mutation CCC,CCT P195P XP_011521889.1

View Full Product Details