Product Details

SNP ID
rs199606157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81666897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCAGCCCCGTGGCCCGGGCTTC[G/T]CAGGTGACTGCGCTGCCCCGCGAGG
Phenotype
MIM: 614271 MIM: 180073
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC137 PubMed Links

Gene Details

Gene
CCDC137
Gene Name
coiled-coil domain containing 137
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199287.2 201 Missense Mutation CGC,CTC R44L NP_954981.1
XM_011524738.2 201 Intron XP_011523040.1
XM_017024573.1 201 Missense Mutation CGC,CTC R60L XP_016880062.1
Gene
OXLD1
Gene Name
oxidoreductase like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039842.2 201 Intron NP_001034931.1
NM_001304994.1 201 Intron NP_001291923.1
NM_001304995.1 201 Intron NP_001291924.1
NM_001304999.1 201 Intron NP_001291928.1
Gene
PDE6G
Gene Name
phosphodiesterase 6G
There are no transcripts associated with this gene.

View Full Product Details