Product Details

SNP ID
rs200219759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74695726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGCCCCCATGGAGGACGCACC[A/G]TGGTGACATATTCCACTTCCACCTG
Phenotype
MIM: 609807 MIM: 609956
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD300LF PubMed Links

Gene Details

Gene
CD300LF
Gene Name
CD300 molecule like family member f
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289082.1 926 UTR 3 NP_001276011.1
NM_001289083.1 926 UTR 3 NP_001276012.1
NM_001289084.1 926 Missense Mutation ACG,ATG T254M NP_001276013.1
NM_001289085.1 926 Missense Mutation ACG,ATG T242M NP_001276014.1
NM_001289086.1 926 UTR 3 NP_001276015.1
NM_001289087.1 926 UTR 3 NP_001276016.1
NM_139018.4 926 Missense Mutation ACG,ATG T239M NP_620587.2
XM_017024212.1 926 Missense Mutation ACG,ATG T258M XP_016879701.1
XM_017024213.1 926 UTR 3 XP_016879702.1
XM_017024214.1 926 UTR 3 XP_016879703.1
XM_017024215.1 926 UTR 3 XP_016879704.1
XM_017024216.1 926 UTR 3 XP_016879705.1
XM_017024217.1 926 UTR 3 XP_016879706.1
Gene
RAB37
Gene Name
RAB37, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006638.2 926 Intron NP_001006639.1
NM_001163989.1 926 Intron NP_001157461.1
NM_001163990.1 926 Intron NP_001157462.1
NM_175738.4 926 Intron NP_783865.1
XM_005257288.4 926 Intron XP_005257345.1

View Full Product Details