Product Details

SNP ID
rs200255480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81120243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTGACGTCGTCGAAGAAGGACAC[G/T]GCCTTCTTCTTGCGTTCCAGGTCCT
Phenotype
MIM: 605276 MIM: 605475 MIM: 614059
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AATK PubMed Links

Gene Details

Gene
AATK
Gene Name
apoptosis associated tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080395.2 3618 Silent Mutation NP_001073864.2
NM_004920.2 3618 Silent Mutation NP_004911.2
XM_006722193.2 3618 Silent Mutation XP_006722256.1
XM_006722194.2 3618 Silent Mutation XP_006722257.1
XM_006722195.2 3618 Silent Mutation XP_006722258.1
XM_006722196.2 3618 Silent Mutation XP_006722259.1
XM_011525509.2 3618 Silent Mutation XP_011523811.1
XM_011525510.2 3618 Silent Mutation XP_011523812.1
XM_011525511.2 3618 Silent Mutation XP_011523813.1
XM_017025421.1 3618 Silent Mutation XP_016880910.1
XM_017025422.1 3618 Silent Mutation XP_016880911.1
Gene
BAIAP2
Gene Name
BAI1 associated protein 2
There are no transcripts associated with this gene.

Gene
MIR3065
Gene Name
microRNA 3065
There are no transcripts associated with this gene.

Gene
MIR338
Gene Name
microRNA 338
There are no transcripts associated with this gene.

Gene
MIR657
Gene Name
microRNA 657
There are no transcripts associated with this gene.

View Full Product Details