Product Details

SNP ID
rs200122136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34270360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTTCAGCCCCCAGGGGCTTGCT[C/T]AGCCAGGTAAGGTCCCTCTCTCCTT
Phenotype
MIM: 158106
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL7 PubMed Links

Gene Details

Gene
CCL7
Gene Name
C-C motif chemokine ligand 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006273.3 145 Nonsense Mutation CAG,TAG Q24* NP_006264.2

View Full Product Details