Product Details

SNP ID
rs199680696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58155241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCTCCAGAAATTCCTGTTCCTT[C/T]TGTTCCTGTTAGTCTATGTTACCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MSX2P1 PubMed Links

Gene Details

Gene
MSX2P1
Gene Name
msh homeobox 2 pseudogene 1
There are no transcripts associated with this gene.

Gene
OR4D1
Gene Name
olfactory receptor family 4 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012374.1 88 Silent Mutation CTG,TTG L30L NP_036506.1

View Full Product Details