Product Details
- SNP ID
-
rs200160703
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40876367 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTCCTTCTGGTAGCTATTTAGAT[A/G]TTTTCTTCCACCTCTTTGACTTCAG
- Phenotype
-
MIM: 601687
MIM: 608218
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT12
PubMed Links
Gene Details
- Gene
- KRT12
- Gene Name
- keratin 12
There are no transcripts associated with this gene.
- Gene
- KRT20
- Gene Name
- keratin 20
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019010.2 |
1327 |
Silent Mutation |
AAC,AAT |
N423N |
NP_061883.1 |
View Full Product Details