Product Details

SNP ID
rs199545653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8075670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGCAATTCCGGATCACCCTCCA[C/T]GGCTGCGTCACTCGGGTAATAATAG
Phenotype
MIM: 603741
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALOX12B PubMed Links

Gene Details

Gene
ALOX12B
Gene Name
arachidonate 12-lipoxygenase, 12R type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139.2 1840 Missense Mutation ATG,GTG M527V NP_001130.1

View Full Product Details