Product Details

SNP ID
rs199616922
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:10521335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCGGCTCTTCACCCGCAGTTTG[C/T]TCACCTGGGACTCAGCAATGTCAGC
Phenotype
MIM: 160730 MIM: 160740
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MYH1 PubMed Links

Gene Details

Gene
MYH1
Gene Name
myosin heavy chain 1
There are no transcripts associated with this gene.

Gene
MYH2
Gene Name
myosin heavy chain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100112.1 5899 Missense Mutation AAC,AGC N1924S NP_001093582.1
NM_017534.5 5899 Missense Mutation AAC,AGC N1924S NP_060004.3
Gene
MYHAS
Gene Name
myosin heavy chain gene cluster antisense RNA
There are no transcripts associated with this gene.

View Full Product Details