Product Details

SNP ID
rs199786137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42538377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCCTGACCCAGGCAGAGATCAA[C/T]GAGTTCTTTACTGGTCCTGCCTTCC
Phenotype
MIM: 609701
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAGLU PubMed Links

Gene Details

Gene
NAGLU
Gene Name
N-acetyl-alpha-glucosaminidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000263.3 647 Silent Mutation AAC,AAT N190N NP_000254.2
XM_006721920.2 647 UTR 5 XP_006721983.1
XM_011524840.1 647 UTR 5 XP_011523142.1
XM_017024686.1 647 UTR 5 XP_016880175.1
XM_017024687.1 647 UTR 5 XP_016880176.1

View Full Product Details