Product Details

SNP ID
rs199855551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43641949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGGCTGTGGAGTCCCCATCCTC[A/C]GGGTCCTGCCCATTGGGGGAGATGG
Phenotype
MIM: 600147
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MEOX1 PubMed Links

Gene Details

Gene
MEOX1
Gene Name
mesenchyme homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040002.1 755 Silent Mutation CCG,CCT P127P NP_001035091.1
NM_004527.3 755 Silent Mutation CCG,CCT P242P NP_004518.1
XM_011524818.1 755 Intron XP_011523120.1

View Full Product Details