Product Details

SNP ID
rs199962301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:27759061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGGCCAGCTGCTGCCGCAGGAT[A/G]TCCTGAACATAGACCTGAAACCAGA
Phenotype
MIM: 163730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOS2 PubMed Links

Gene Details

Gene
NOS2
Gene Name
nitric oxide synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000625.4 3035 Silent Mutation GAC,GAT D1058D NP_000616.3
XM_011524859.2 3035 Silent Mutation GAC,GAT D1058D XP_011523161.1
XM_011524860.2 3035 Silent Mutation GAC,GAT D1057D XP_011523162.1
XM_011524861.1 3035 Silent Mutation GAC,GAT D1034D XP_011523163.1
XM_011524862.1 3035 Silent Mutation GAC,GAT D836D XP_011523164.1

View Full Product Details