Product Details

SNP ID
rs200519924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32021878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTACCTCCAGAACTCCGGGTGAA[C/T]GCAGATGAGCCTCCAGGGCCTCCTG
Phenotype
MIM: 616558
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC37B PubMed Links

Gene Details

Gene
LRRC37B
Gene Name
leucine rich repeat containing 37B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321350.1 937 Silent Mutation AAC,AAT N162N NP_001308279.1
NM_052888.2 937 Intron NP_443120.2

View Full Product Details