Product Details

SNP ID
rs200134501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2033508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGCCTTATATCCATTCTAGGTC[A/G]GGCCCCTCGGGGCCGCGTGGCCAAT
Phenotype
MIM: 603527 MIM: 607896 MIM: 610461
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPH1 PubMed Links

Gene Details

Gene
DPH1
Gene Name
diphthamide biosynthesis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001383.3 98 Missense Mutation CAG,CGG Q27R NP_001374.3
Gene
OVCA2
Gene Name
ovarian tumor suppressor candidate 2
There are no transcripts associated with this gene.

Gene
RTN4RL1
Gene Name
reticulon 4 receptor like 1
There are no transcripts associated with this gene.

View Full Product Details