Product Details

SNP ID
rs199900544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47744303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACGCTTCCAACACGCATATCTTT[A/G]CTTTCCAAGAAACCCAGTTCATTGC
Phenotype
MIM: 604895
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBX21 PubMed Links

Gene Details

Gene
TBX21
Gene Name
T-box 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013351.1 1155 Missense Mutation ACT,GCT T293A NP_037483.1
XM_011524698.2 1155 Missense Mutation ACT,GCT T314A XP_011523000.1

View Full Product Details