Product Details
- SNP ID
-
rs200443029
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:48529554 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGTGGCTAGGTTCAGTTCAGGAG[G/T]TGACAGAGCTGGGTGAGGCTTCCGG
- Phenotype
-
MIM: 142968
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HOXB1
PubMed Links
Gene Details
- Gene
- HOXB1
- Gene Name
- homeobox B1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002144.3 |
905 |
Missense Mutation |
AAC,ACC |
N300T |
NP_002135.2 |
View Full Product Details