Product Details
- SNP ID
-
rs200443432
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41460135 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGTAGCGGCCCTGGGGGCAGGGT[G/T]AGCAAGGCATGCCAACAGTGCGTGG
- Phenotype
-
MIM: 602760
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT32
PubMed Links
Gene Details
- Gene
- KRT32
- Gene Name
- keratin 32
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002278.3 |
1383 |
Nonsense Mutation |
TAA,TCA |
*441S |
NP_002269.3 |
View Full Product Details