Product Details

SNP ID
rs200562682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44689651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGCTCCATAAACGGCTCGGTCCA[C/T]TCCCAGTGCCTCCAACCGTCCGTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC43 PubMed Links

Gene Details

Gene
CCDC43
Gene Name
coiled-coil domain containing 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099225.1 147 Missense Mutation ATG,GTG M35V NP_001092695.1
NM_144609.2 147 Missense Mutation ATG,GTG M35V NP_653210.2

View Full Product Details