Product Details
- SNP ID
-
rs199592890
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:16939796 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGGCTGCCGGCTTCCATCGCGTG[A/G]TCTGCAGAGGCGAGAGTGGAGGGCG
- Phenotype
-
MIM: 604907
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TNFRSF13B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11652811] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TNFRSF13B
- Gene Name
- TNF receptor superfamily member 13B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_012452.2 |
646 |
Silent Mutation |
GAC,GAT |
D211D |
NP_036584.1 |
View Full Product Details