Product Details
- SNP ID
-
rs202226873
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41363874 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTTGCAAGGCCCACAGCGGGAAC[G/T]AGGACCACAAGGATTGGTGACACAG
- Phenotype
-
MIM: 602762
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT33B
PubMed Links
Gene Details
- Gene
- KRT33B
- Gene Name
- keratin 33B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002279.4 |
1227 |
Missense Mutation |
AGT,CGT |
S393R |
NP_002270.1 |
View Full Product Details