Product Details

SNP ID
rs200782457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44349427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATCATTTTCCCTGTCTTTCTAGG[A/T]CAAATGGCCCACAACACTGAGCAGG
Phenotype
MIM: 138945
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM171A2 PubMed Links

Gene Details

Gene
FAM171A2
Gene Name
family with sequence similarity 171 member A2
There are no transcripts associated with this gene.

Gene
GRN
Gene Name
granulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002087.3 396 Missense Mutation GAC,GTC D47V NP_002078.1
XM_005257253.1 396 Missense Mutation GAC,GTC D47V XP_005257310.1

View Full Product Details