Product Details

SNP ID
rs201506755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:19380806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGAACTGTGAGCTCAAGATTGG[A/T]GACTTTGGTATGGCTCGTGGCCTGT
Phenotype
MIM: 614144 MIM: 602521 MIM: 600596
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
B9D1 PubMed Links

Gene Details

Gene
B9D1
Gene Name
B9 domain containing 1
There are no transcripts associated with this gene.

Gene
MAPK7
Gene Name
mitogen-activated protein kinase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002749.3 1001 Silent Mutation GGA,GGT G199G NP_002740.2
NM_139032.2 1001 Silent Mutation GGA,GGT G60G NP_620601.1
NM_139033.2 1001 Silent Mutation GGA,GGT G199G NP_620602.2
NM_139034.2 1001 Silent Mutation GGA,GGT G199G NP_620603.2
XM_006721557.3 1001 Silent Mutation GGA,GGT G205G XP_006721620.1
XM_006721558.3 1001 Silent Mutation GGA,GGT G205G XP_006721621.1
XM_006721559.3 1001 Silent Mutation GGA,GGT G205G XP_006721622.1
XM_011523957.2 1001 Silent Mutation GGA,GGT G60G XP_011522259.1
Gene
MFAP4
Gene Name
microfibrillar associated protein 4
There are no transcripts associated with this gene.

View Full Product Details