Product Details
- SNP ID
-
rs201541265
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:72122915 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCCATCTTCAAGGCGCTGCAGGCC[A/G]ACTCGCCACACTCCTCCTCCGGCAT
- Phenotype
-
MIM: 608160
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SOX9
PubMed Links
Gene Details
- Gene
- SOX9
- Gene Name
- SRY-box 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000346.3 |
1000 |
Missense Mutation |
AAC,GAC |
N210D |
NP_000337.1 |
- Gene
- SOX9-AS1
- Gene Name
- SOX9 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details