Product Details

SNP ID
rs200961470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58243004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGAGGGTCCTTCTCCATCTCCCA[C/G]CCCTCCTGGCTTCCCTCATCTTGCT
Phenotype
MIM: 150205
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LPO PubMed Links

Gene Details

Gene
LPO
Gene Name
lactoperoxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160102.1 217 Missense Mutation CCC,GCC P9A NP_001153574.1
NM_006151.2 217 Missense Mutation CCC,GCC P9A NP_006142.1
XM_011524808.2 217 Missense Mutation CCC,GCC P9A XP_011523110.1
XM_011524810.2 217 Intron XP_011523112.1

View Full Product Details