Product Details

SNP ID
rs200652567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28910191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATTCGCACACGTAGAGGGGAGA[G/T]CTGATGATGTGGTGACAGGTGTGTA
Phenotype
MIM: 616157
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DHRS13 PubMed Links

Gene Details

Gene
DHRS13
Gene Name
dehydrogenase/reductase 13
There are no transcripts associated with this gene.

Gene
PHF12
Gene Name
PHD finger protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033561.1 2924 Intron NP_001028733.1
NM_001290131.1 2924 Missense Mutation AGA,AGC R798S NP_001277060.1
NM_020889.2 2924 Intron NP_065940.1
XM_005258015.3 2924 Intron XP_005258072.1
XM_005258016.3 2924 Intron XP_005258073.1
XM_011525078.2 2924 Intron XP_011523380.1
XM_011525079.2 2924 Intron XP_011523381.1
XM_011525082.2 2924 Intron XP_011523384.1
XM_011525083.2 2924 Intron XP_011523385.1
XM_017024903.1 2924 Intron XP_016880392.1
XM_017024904.1 2924 Intron XP_016880393.1

View Full Product Details