Product Details
- SNP ID
-
rs201648544
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:45396207 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCAGGGGTTGGGGACGGGCCTCAC[C/G]GTGGTCCACCTTATAGCGTAGCTTC
- Phenotype
-
MIM: 610591
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGAP27
PubMed Links
Gene Details
- Gene
- ARHGAP27
- Gene Name
- Rho GTPase activating protein 27
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282290.1 |
2651 |
Missense Mutation |
CAT,GAT |
H751D |
NP_001269219.1 |
NM_174919.3 |
2651 |
Intron |
|
|
NP_777579.2 |
NM_199282.2 |
2651 |
Missense Mutation |
CAT,GAT |
H410D |
NP_954976.1 |
XM_005257128.3 |
2651 |
Missense Mutation |
CAT,GAT |
H529D |
XP_005257185.1 |
XM_006721745.2 |
2651 |
Missense Mutation |
CAT,GAT |
H751D |
XP_006721808.1 |
XM_006721747.2 |
2651 |
Missense Mutation |
CAT,GAT |
H729D |
XP_006721810.1 |
XM_006721750.3 |
2651 |
Intron |
|
|
XP_006721813.1 |
XM_011524468.1 |
2651 |
Missense Mutation |
CAT,GAT |
H784D |
XP_011522770.1 |
XM_011524469.1 |
2651 |
Missense Mutation |
CAT,GAT |
H762D |
XP_011522771.1 |
XM_011524470.2 |
2651 |
Missense Mutation |
CAT,GAT |
H784D |
XP_011522772.1 |
XM_011524471.1 |
2651 |
Missense Mutation |
CAT,GAT |
H727D |
XP_011522773.1 |
XM_011524472.2 |
2651 |
Missense Mutation |
CAT,GAT |
H584D |
XP_011522774.1 |
XM_011524473.2 |
2651 |
Intron |
|
|
XP_011522775.1 |
XM_011524474.1 |
2651 |
Missense Mutation |
CAT,GAT |
H443D |
XP_011522776.1 |
XM_017024311.1 |
2651 |
Missense Mutation |
CAT,GAT |
H784D |
XP_016879800.1 |
XM_017024312.1 |
2651 |
Missense Mutation |
CAT,GAT |
H551D |
XP_016879801.1 |
XM_017024313.1 |
2651 |
Intron |
|
|
XP_016879802.1 |
View Full Product Details